RIE321609182 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-yl)-hexan-2-one

  • MDL number MFCD18433495
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9501

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9501
Availability details for
Quantity
  • Availability
  • Add to Cart