RIE321609183 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-yl)-heptan-2-one

  • MDL number MFCD18433496
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9502

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9502
Availability details for
Quantity
  • Availability
  • Add to Cart