RIE321609189 Rieke Metals, Inc.

(1,3-Dioxolan-2-yl)-acetic acid propyl ester

  • MDL number MFCD18433504
  • Molecular Weight : 174.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9510

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9510
Availability details for
Quantity
  • Availability
  • Add to Cart