RIE321609190 Rieke Metals, Inc.

(1,3-Dioxolan-2-yl)-acetic acid butyl ester

  • MDL number MFCD18433505
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9511

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9511
Availability details for
Quantity
  • Availability
  • Add to Cart