RIE321609193 Rieke Metals, Inc.

6-Chloro-1-(1,3-dioxolan-2-yl)-hexan-2-one

  • MDL number MFCD18433508
  • Molecular Weight : 206.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9514

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9514
Availability details for
Quantity
  • Availability
  • Add to Cart