RIE321609196 Rieke Metals, Inc.

1-Cyclopentyl-2-(1,3-dioxolan-2-yl)-ethanone

  • MDL number MFCD18433511
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9517

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9517
Availability details for
Quantity
  • Availability
  • Add to Cart