RIE321609199 Rieke Metals, Inc.

1-Cyclohexyl-3-(1,3-dioxolan-2-yl)-propan-2-one

  • MDL number MFCD18433514
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9520

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9520
Availability details for
Quantity
  • Availability
  • Add to Cart