RIE321609200 Rieke Metals, Inc.

4-Cyclohexyl-1-(1,3-dioxolan-2-yl)-butan-2-one

  • MDL number MFCD18433515
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9521

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9521
Availability details for
Quantity
  • Availability
  • Add to Cart