RIE321609205 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-yl)-3-ethyl-pentan-2-one

  • MDL number MFCD18433521
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9527

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9527
Availability details for
Quantity
  • Availability
  • Add to Cart