RIE321609219 Rieke Metals, Inc.

3-Fluoro-2-methylphenyl magnesium bromide

  • MDL number MFCD01311483
  • Cas Number : 263355-05-1
  • Molecular Weight : 213.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3235

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3235
Availability details for
Quantity
  • Availability
  • Add to Cart