RIE321609225 Rieke Metals, Inc.

2-(1,3-Dioxolan-2-yl)-1-phenyl-ethanone

  • MDL number MFCD00156699
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9001

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9001
Availability details for
Quantity
  • Availability
  • Add to Cart