RIE321609231 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-yl)-3,3-dimethyl-butan-2-one

  • MDL number MFCD18433517
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9523

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9523
Availability details for
Quantity
  • Availability
  • Add to Cart