RIE341038355 Rieke Metals, Inc.

2-Pyridinylmethyl zinc chloride

  • MDL number MFCD19687266
  • Cas Number : 165334-81-6
  • Molecular Weight : 192.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/2306

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2306
Availability details for
Quantity
  • Availability
  • Add to Cart