RIE370208429 Rieke Metals, Inc.

3-Trifluoromethylphenethylzinc bromide

  • MDL number MFCD25562969
  • Molecular Weight : 318.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2314

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2314
Availability details for
Quantity
  • Availability
  • Add to Cart