RIE370208430 Rieke Metals, Inc.

3,4-Dichlorophenethylmagnesium bromide

  • MDL number MFCD25562972
  • Molecular Weight : 278.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3239

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3239
Availability details for
Quantity
  • Availability
  • Add to Cart