RIE370208431 Rieke Metals, Inc.

5-Chloro-2-methoxyphenylmagnesium bromide

  • MDL number MFCD11113495
  • Cas Number : 419535-75-4
  • Molecular Weight : 245.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3238

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3238
Availability details for
Quantity
  • Availability
  • Add to Cart