RIE458978982 Rieke Metals, Inc.

6-Fluoro-2,3'-bipyridine

  • MDL number MFCD15475739
  • Cas Number : 1214335-26-8
  • Molecular Weight : 174.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/11178

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11178
Availability details for
Quantity
  • Availability
  • Add to Cart