RIE458978983 Rieke Metals, Inc.

4-Chloro-2-methoxyphenylzinc bromide

  • MDL number MFCD22684792
  • Molecular Weight : 286.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2337

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2337
Availability details for
Quantity
  • Availability
  • Add to Cart