RIE458978990 Rieke Metals, Inc.

3-iso-Propylphenylmagnesium bromide

  • MDL number MFCD06201614
  • Molecular Weight : 223.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3245

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3245
Availability details for
Quantity
  • Availability
  • Add to Cart