RIE515935248 Rieke Metals, Inc.

4-Fluorophenylzinc iodide

  • Cas Number : 674346-88-4
  • Molecular Weight : 287.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/2361

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2361
Availability details for
Quantity
  • Availability
  • Add to Cart