RIE515935258 Rieke Metals, Inc.

2-tert-Butoxy-2-oxoethylzinc chloride

  • MDL number MFCD00671972
  • Cas Number : 321745-86-2
  • Molecular Weight : 215.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/2355

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2355
Availability details for
Quantity
  • Availability
  • Add to Cart