RIEH0429B91C Rieke Metals, Inc.

4-(4-Bromo-3-fluorophenyl)-1-butene

  • Cas Number : 1442114-00-2
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-1071b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1071b
Availability details for
Quantity
  • Availability
  • Add to Cart