RIEH0429B91E Rieke Metals, Inc.

2-Bromo-4-(4-bromo-3-fluorophenyl)-1-butene

  • Molecular Weight : 307.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10005-1073B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1073B
Availability details for
Quantity
  • Availability
  • Add to Cart