RIEH0429B91F Rieke Metals, Inc.

4-(4-Bromo-3-fluorophenyl)-2-methyl-1-butene

  • Molecular Weight : 243.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10005-1074B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1074B
Availability details for
Quantity
  • Availability
  • Add to Cart