RIEH0429B920 Rieke Metals, Inc.

4-(3-Chloro-5-fluorophenyl)-1-butene

  • Molecular Weight : 184.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10005-1101B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1101B
Availability details for
Quantity
  • Availability
  • Add to Cart