RIEH0429B929 Rieke Metals, Inc.

2-Bromo-4-(4-fluoro-2-methylphenyl)-1-butene

  • Molecular Weight : 243.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-1163b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1163b
Availability details for
Quantity
  • Availability
  • Add to Cart