RIEH0429B92E Rieke Metals, Inc.

3-(5-Bromo-2-fluorophenyl)-2-chloro-1-propene

  • Molecular Weight : 249.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-1172a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1172a
Availability details for
Quantity
  • Availability
  • Add to Cart