RIEH0429B972 Rieke Metals, Inc.

5-(3,4-Dichlorophenyl)-2-methyl-1-pentene

  • Molecular Weight : 229.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-684c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-684c
Availability details for
Quantity
  • Availability
  • Add to Cart