RIEH0429B974 Rieke Metals, Inc.

2-Chloro-3-(2,5-difluorophenyl)-1-propene

  • Molecular Weight : 188.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-712a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-712a
Availability details for
Quantity
  • Availability
  • Add to Cart