RIEH0429B9C2 Rieke Metals, Inc.

2-(Trifluoromethoxy)phenylmagnesium bromide

  • MDL number MFCD18253607
  • Cas Number : 944470-93-3
  • Molecular Weight : 265.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3322

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3322
Availability details for
Quantity
  • Availability
  • Add to Cart