SY3371475593 Synthonix Corporation

7-Azabicyclo[2.2.1]heptane

  • MDL number MFCD13624216
  • Cas Number : 279-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0698.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0698.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart