SY3371475606 Synthonix Corporation

Phytosphingosine

  • MDL number MFCD02259274
  • Cas Number : 554-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/0929.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0929.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart