SY3371475623 Synthonix Corporation

(5-Amino-2-hydroxymethylphenyl)boronic acid HCl

  • MDL number MFCD04115645
  • Cas Number : 117098-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/1210.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1210.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart