SY3371475625 Synthonix Corporation

3-Aminomethylphenylboronic acid hydrochloride

  • MDL number MFCD02093050
  • Cas Number : 352525-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/1260.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1260.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart