SY3371475660 Synthonix Corporation

4-Amino-1-(methanesulfonyl)piperidine

  • MDL number MFCD06805736
  • Cas Number : 402927-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1637.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1637.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart