SY3371475673 Synthonix Corporation

2-(Aminoethyl)-5-bromopyridine

  • MDL number MFCD08449700
  • Cas Number : 691872-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1731.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1731.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart