SY3371475680 Synthonix Corporation

6-Amino-pyridazine-3-carbonitrile

  • MDL number MFCD09743651
  • Cas Number : 340759-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1858.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1858.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart