SY3371475692 Synthonix Corporation

3-Amino-2-ethoxycarbonylpyrrole hydrochloride

  • MDL number MFCD04038917
  • Cas Number : 252932-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/2113.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2113.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart