SY3371475719 Synthonix Corporation

6-Amino-2-methyl-2H-indazole

  • MDL number MFCD00464866
  • Cas Number : 50593-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/2918.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2918.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart