SY3371475725 Synthonix Corporation

2-[(2-Aminoethylthio)methyl]furan

  • MDL number MFCD01075029
  • Cas Number : 36415-21-1
  • Molecular Weight : 157.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(2-furylmethyl)sulfanyl]ethylamine
Inchi Code 1S/C7H11NOS/c8-3-5-10-6-7-2-1-4-9-7/h1-2,4H,3,5-6,8H2
Inchi Key VSPDYEHKAMKDNW-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3034.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3034.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart