SY3371475744 Synthonix Corporation

Methyl 2-amino-5-ethylthiazole-4-carboxylate

  • MDL number MFCD00728625
  • Cas Number : 28942-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3487.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/3487.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart