SY3371475745 Synthonix Corporation

2-Aminothiazole-5-carboxylic acid

  • MDL number MFCD06203554
  • Cas Number : 40283-46-3
  • Molecular Weight : 144.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3492.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3492.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart