SY3371475793 Synthonix Corporation

1-Aminopyrrole-2-carboxamide

  • Cas Number : 159326-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/5122.html

Safety Information

MSDS https://synthonix.com/5122.html
Availability details for
Quantity
  • Availability
  • Add to Cart