SY3371475819 Synthonix Corporation

2-Aminomethyl-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

  • MDL number MFCD12198554
  • Cas Number : 1160247-15-3
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5996.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5996.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart