SY3371475829 Synthonix Corporation

3-Amino-3-methyl-N-benzylazetidine

  • MDL number MFCD18839235
  • Cas Number : 943437-96-5
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6073.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6073.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart