SY3371475832 Synthonix Corporation

2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoro acetate

  • MDL number MFCD19982773
  • Cas Number : 1523572-02-2
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6139.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6139.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart