SY3371475843 Synthonix Corporation

1-(Azetidin-3-yl)-4,4-difluoropiperidine ditrifluoroacetic acid salt

  • MDL number MFCD23152699
  • Cas Number : 1257293-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6315.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6315.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart