SY3371475846 Synthonix Corporation

1-(Azetidin-3-yl)pyrrolidin-2-one trifluoro acetate

  • MDL number MFCD22628911
  • Cas Number : 1257294-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6322.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6322.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart