SY3371475847 Synthonix Corporation

1-(Azetidin-3-yl)-3,3-difluoropyrrolidine ditrifluoroacetate

  • MDL number MFCD22628910
  • Cas Number : 1706462-59-0
  • Molecular Weight : 162.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6325.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6325.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart