SY3371475856 Synthonix Corporation

2-Azabicyclo[2.2.2]octane-2-carboxylic acid, 5-oxo-,1,1-dimethylethyl ester

  • MDL number MFCD17016701
  • Cas Number : 617714-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/6584.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6584.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart